missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-GAPDH (High Potency), MP Biomedicals™

Catalog No. MP08L100030
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08L100030 Supplier MP Biomedicals Inc Supplier No. 08L100030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

  • Mouse anti-GAPDH mAb (High Potency) offers high specificity and potency and consistency against GAPDH expressed by human, mouse, and rat species, which has been experimentally validated in Western blotting (WB), Immunohistochemistry (IHC), and Immunofluorescence (IF) assays.
  • This gene encodes a member of the glyceraldehyde-3-phosphate dehydrogenase protein family. The encoded protein has been identified as a moonlighting protein based on its ability to perform mechanistically distinct functions.
  • The product of this gene catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD). The encoded protein has additionally been identified to have uracil DNA glycosylase activity in the nucleus.

Specifications

Antigen GAPDH
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Conjugate Unconjugated
Formulation In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol; pH 7.3 to 7.4
Host Species Mouse
Immunogen Full-length recombinant protein of human GAPDH
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human, Mouse, Rat
Antibody Molecular Weight Observed at 36 kDa
Form Purified
Isotype IgG
Show More Show Less

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.