missing translation for 'onlineSavingsMsg'
Learn More

Mouse Anti-Chicken IgY Antibody [4A4A10] (HRP), ProSci

Catalog No. 16917039
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16917039 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16917039 Supplier ProSci Llc Supplier No. 16090100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Secondary Antibody

Mouse Anti-Chicken IgY Antibody specifically detects Mouse Anti-Chicken IgY in Chicken Samples. It is validated for ELISA, Western Blot, Immunohistochemistry.

Specifications

Antigen Mouse Anti-Chicken IgY
Applications ELISA, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 4A4A10
Concentration 1 mg/mL
Conjugate HRP
Dilution Immunoblotting: 1/5,000 - 1/50,000, ELISA: 1/5,000 - 1/50,000, Immunohistochemistry: 1/100 - 1/1,000
Formulation PBS pH 7.2, 1% BSA, 0.02% Thimerosal
Host Species Mouse
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No cross reactivity with immunoglobulins from other species.
Target Species Chicken
Content And Storage Store at 2-8°C. The product is stable for about 4-6 weeks at 4°C as an undiluted liquid. For extended storage, aliquot and freeze the product at -70°C or below. Avoid repeated freezing and thawing. Or alternatively, add an equal volume of glycerol for a final concentration of 50% and store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less