missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Canine IgG2 Heavy Chain Secondary Antibody (CA4F1) [Janelia Fluor 525], R&D Systems™

Catalog No. NB784621
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB784621 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB784621 Supplier Bio-Techne Supplier No. NBP268409JF525

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG2 Heavy Chain Monoclonal antibody specifically detects IgG2 Heavy Chain in Canine samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen IgG2 Heavy Chain
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone CA4F1
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Canine IgG
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Canine
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less