missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Bovine IgM (H+L) Secondary Antibody (IL-A30) [Janelia Fluor 669], R&D Systems™

Catalog No. NB781284
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB781284 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB781284 Supplier Bio-Techne Supplier No. NBP268419JF669

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgM (H+L) Monoclonal antibody specifically detects IgM (H+L) in Bovine samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Radioimmune Assays

Specifications

Antigen IgM (H+L)
Applications Flow Cytometry, ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone IL-A30
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Bovine peripheral blood mononuclear cells depleted of T lymphocytes
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less