missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Bovine IgG2 Heavy Chain Secondary Antibody (IL-A73) [Janelia Fluor 669], R&D Systems™
Description
IgG2 Heavy Chain Monoclonal antibody specifically detects IgG2 Heavy Chain in Bovine samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation
Specifications
Specifications
| Antigen | IgG2 Heavy Chain |
| Applications | Flow Cytometry, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | IL-A73 |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Purified bovine IgG2 |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |