missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Bovine IgG1 Secondary Antibody, HRP

Catalog No. PIMA516736
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516736 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516736 Supplier Invitrogen™ Supplier No. MA516736
  • ONLINE EXCLUSIVE PRICE
    $897.89
    $522.00 / Each

    Save $375.89

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Secondary Antibody

Mouse anti Bovine IgG1 antibody, clone IL-A60 recognizes bovine IgG1. Mouse anti Bovine IgG1 antibody, clone IL-A60 immunoprecipitates a protein band of 55-59 kDa, consistent with the heavy chain of bovine IgG1 (Campbell et al. 1998).

Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Bovine IgG1
Applications ELISA
Classification Monoclonal
Clone IL-A60
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 0.01% thimerosal
Host Species Mouse
Immunogen Purified bovine Immunoglobulin.
Purification Method Protein A
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less