missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Bovine IgG (H+L) Secondary Antibody (IL-A2) [Janelia Fluor 525], R&D Systems™

Catalog No. NB784619
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB784619 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB784619 Supplier Bio-Techne Supplier No. NBP268413JF525

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG (H+L) Monoclonal antibody specifically detects IgG (H+L) in Bovine samples. It is validated for ELISA, Immunoprecipitation

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone IL-A2
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Purified bovine IgG
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less