missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Bovine IgA Heavy Chain Secondary Antibody (IL-A71) [Janelia Fluor 525], R&D Systems™

Catalog No. NB784622
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB784622 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB784622 Supplier Bio-Techne Supplier No. NBP268410JF525

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgA Heavy Chain Monoclonal antibody specifically detects IgA Heavy Chain in Bovine samples. It is validated for ELISA, Immunoprecipitation

Specifications

Antigen IgA Heavy Chain
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone IL-A71
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Purified bovine Immunoglobulin
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less