missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Bovine Ig Mix Secondary Antibody
Mouse Monoclonal Secondary Antibody
Supplier: Invitrogen™ SA110637
Description
SA1-10637 detects bovine Immunoglobulin. Does not cross react with canine, equine, feline, rabbit, ovine, porcine, donkey, guinea pig, goat, or human. SA1-10637 has been successfully used in ELISA applications. The SA1-10637 immunogen is purified bovine immunoglobulin.
Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Bovine Ig Mix | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Purified bovine immunoglobulin. | |
| 500 μg | |
| Secondary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| BIG10-101.6 | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Bovine | |
| Secondary Antibody | |
| IgG1 |