missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Morpholinoacetonitrile 98.0+%, TCI America™
Supplier: TCI America M109525G
Specifications
| Morpholinoacetonitrile | |
| 62°C | |
| 124°C | |
| MFCD00023372 | |
| 3439 | |
| OOSOCAXREAGIGA-UHFFFAOYSA-N | |
| 2-morpholin-4-ylacetonitrile | |
| 22055 | |
| ≥98.0% (GC,T) |
| 5807-02-3 | |
| White | |
| C6H10N2O | |
| 25 g | |
| N-Cyanomethylmorpholine | |
| C1COCCN1CC#N | |
| 126.16 | |
| 126.16 | |
| Crystalline Powder |
Chemical Identifiers
| 5807-02-3 | |
| 126.16 | |
| OOSOCAXREAGIGA-UHFFFAOYSA-N | |
| 22055 | |
| C1COCCN1CC#N |
| C6H10N2O | |
| MFCD00023372 | |
| N-Cyanomethylmorpholine | |
| 2-morpholin-4-ylacetonitrile |
Safety and Handling
RTECSNumber : QD6650000
TSCA : No