accessibility menu, dialog, popup

UserName

CAS RN 5807-02-3

IUPAC Name: 2-(morpholin-4-yl)acetonitrile
Synonyms: 4-morpholineacetonitrile
Molecular Weight (g/mol): 126.16
Molecular Formula: C6H10N2O
InChi Key: OOSOCAXREAGIGA-UHFFFAOYSA-N
SMILES: N#CCN1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results