missing translation for 'onlineSavingsMsg'
Learn More

Morpholinoacetonitrile 98.0+%, TCI America™

Catalog No. p-7132535
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10955G 5 g
M109525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10955G Supplier TCI America Supplier No. M10955G
Only null left

Chemical Identifiers

CAS 5807-02-3
Molecular Formula C6H10N2O
Molecular Weight (g/mol) 126.16
MDL Number MFCD00023372
InChI Key OOSOCAXREAGIGA-UHFFFAOYSA-N
Synonym N-Cyanomethylmorpholine
PubChem CID 22055
IUPAC Name 2-morpholin-4-ylacetonitrile
SMILES C1COCCN1CC#N

Specifications

Melting Point 62°C
Color White
Boiling Point 124°C
Quantity 5 g
UN Number 3439
Formula Weight 126.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Morpholinoacetonitrile
RTECSNumber QD6650000
TSCA No