missing translation for 'onlineSavingsMsg'
Learn More

Morpholine Standard, 1000 ppm C4H9NO, Ricca Chemical

Catalog No. 521816
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
521816 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 521816 Supplier Ricca Chemical Company Supplier No. 521816

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-91-8 , 7732-18-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1000ppm
CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Physical Form Liquid
Color Colorless
pH Alkaline
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Morpholine Standard
Show More Show Less