missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, ACS, 99%, Spectrum™ Chemical

$553.75 - $553.75

Chemical Identifiers

CAS 110-91-8
Molecular Weight (g/mol) 87.12 g/mol
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611826
View Documents
Spectrum Chemical Mfg Cor
M1340500MLGL
500 mL Steel Can
Each for $553.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

110-91-8
87.12 g/mol

Specifications

Steel Can
1
500 mL
ACS
110-91-8
87.12 g/mol
99%
Morpholine
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.