missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, ACS, 99%, Spectrum™ Chemical

Catalog No. 18611826
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Steel Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611826 500 mL Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18611826 Supplier Spectrum Chemical Mfg Cor Supplier No. M1340500MLGL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-91-8
Molecular Weight (g/mol) 87.12 g/mol

Specifications

Packaging Steel Can
CAS Max % 1
Quantity 500 mL
Percent Purity 99%
Grade ACS
Chemical Name or Material Morpholine

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.