missing translation for 'onlineSavingsMsg'
Learn More

MOPSO sodium salt

Catalog No. AC462140250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462140250 25 g
AC462141000 100 g
AC462145000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC462140250 Supplier Thermo Scientific Chemicals Supplier No. 462140250
Only null left

CAS: 79803-73-9 | C7H14NNaO5S | 247.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79803-73-9
Molecular Formula C7H14NNaO5S
Molecular Weight (g/mol) 247.24
InChI Key WSFQLUVWDKCYSW-UHFFFAOYNA-M
IUPAC Name sodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate
SMILES [Na+].OC(CN1CCOCC1)CS([O-])(=O)=O

Specifications

Quantity 25 g
Solubility Information Solubility aq. soln.: 500 g/L
Formula Weight 247.24
Chemical Name or Material MOPSO sodium salt
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only