missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MOPS, Sodium, ULTROL™ Grade, Calbiochem™,
A zwitterionic buffer useful in the pH range of 6.5 to 7.9
$207.00 - $1020.00
Chemical Identifiers
| CAS | 71119-22-7 |
|---|---|
| Molecular Formula | C7H14NNaO4S |
| Molecular Weight (g/mol) | 231.242 |
| InChI Key | MWEMXEWFLIDTSJ-UHFFFAOYSA-M |
| Synonym | 3-(N-Morpholino)propanesulfonic Acid, Na |
| PubChem CID | 3859613 |
| IUPAC Name | sodium;3-morpholin-4-ylpropane-1-sulfonate |
| SMILES | C1COCCN1CCCS(=O)(=O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
475899100GM
|
MilliporeSigma™
475899100GM |
100 g |
Each for $207.00
|
|
|||||
|
4758991KG
|
MilliporeSigma™
4758991KG |
1 kg |
Each for $1,020.00
|
|
|||||
Chemical Identifiers
| 71119-22-7 | |
| 231.242 | |
| 3-(N-Morpholino)propanesulfonic Acid, Na | |
| sodium;3-morpholin-4-ylpropane-1-sulfonate |
| C7H14NNaO4S | |
| MWEMXEWFLIDTSJ-UHFFFAOYSA-M | |
| 3859613 | |
| C1COCCN1CCCS(=O)(=O)[O-].[Na+] |
Specifications
| 71119-22-7 | |
| ≥98.0% | |
| 3-(N-Morpholino)propanesulfonic Acid, Na | |
| MWEMXEWFLIDTSJ-UHFFFAOYSA-M | |
| sodium;3-morpholin-4-ylpropane-1-sulfonate | |
| 3859613 | |
| by titration (dry basis) |
| 100 g | |
| C7H14NNaO4S | |
| Soluble in cold water | |
| C1COCCN1CCCS(=O)(=O)[O-].[Na+] | |
| 231.242 | |
| 231.2 | |
| zwitterionic buffer |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).