missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MOPS, Sodium, ULTROL™ Grade, Calbiochem™,

Catalog No. 475899100GM
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
475899100GM 100 g
4758991KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 475899100GM Supplier MilliporeSigma™ Supplier No. 475899100GM
Only null left

A zwitterionic buffer useful in the pH range of 6.5 to 7.9

Chemical Identifiers

CAS 71119-22-7
Molecular Formula C7H14NNaO4S
Molecular Weight (g/mol) 231.242
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
Synonym 3-(N-Morpholino)propanesulfonic Acid, Na
PubChem CID 3859613
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate
SMILES C1COCCN1CCCS(=O)(=O)[O-].[Na+]

Specifications

Quantity 100 g
Assay Percent Range ≥98.0%
Solubility Information Soluble in cold water
Formula Weight 231.2
Grade by titration (dry basis)
Chemical Name or Material zwitterionic buffer
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).