missing translation for 'onlineSavingsMsg'
Learn More

MOPS sodium salt, 98%

Catalog No. p-169977
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC352590250 25 g Glass bottle
AC352591000 100 g Glass bottle
AC352590010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC352590250 Supplier Thermo Scientific Chemicals Supplier No. 352590250
Only null left

CAS: 71119-22-7 | C7H14NNaO4S | 231.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

Molecular Formula C7H14NNaO4S
Molecular Weight (g/mol) 231.242
MDL Number MFCD00064350
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
Synonym 3-(4-Morpholino)propanesulfonic acid sodium salt
PubChem CID 3859613
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate
SMILES C1COCCN1CCCS(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 97.5
Color White
CAS Max % 100.0
Physical Form Powder
Quantity 25 g
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 231.25
Percent Purity 98%
Chemical Name or Material MOPS sodium salt, 98%
Show More Show Less

RUO – Research Use Only