missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MOPS SDS Running Buffer for Bis-Tris Gels, Powder

Catalog No. MP08W00025
Change view
Click to view available options
Quantity:
5 Pouches
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08W00025 5 Pouches
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08W00025 Supplier MP Biomedicals™ Supplier No. 08W00025
Only null left

Powder MOPS buffer salts to make PAGE buffer.

MOPS SDS Running Buffer for Bis-Tris Gels in Powder (One Pack makes up 1 L)

Specifications

Physical Form Powder
Quantity 5 Pouches

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.