missing translation for 'onlineSavingsMsg'
Learn More

MOPS, MP Biomedicals™

Catalog No. p-4404455
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10237083 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10237083 Supplier MP Biomedicals Inc Supplier No. 0210237083
Only null left

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-morpholino)propanesulfonic acid, 4-morpholinepropanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 100.4%
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
Formula Weight 209.3
Chemical Name or Material MOPS
EINECSNumber 214-478-5