missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MOPS, Free Acid, ULTROL™ Grade, Calbiochem™,

Catalog No. 475898100GM
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
475898100GM 100 g
475898500GM 500 g
4758981KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 475898100GM Supplier MilliporeSigma™ Supplier No. 475898100GM

A zwitterionic buffer useful in the pH range of 6.5 to 7.9

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-Morpholino)propanesulfonic Acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Quantity 100 g
Assay Percent Range ≥99.0%
Solubility Information Soluble in water
Formula Weight 209.3
Grade by titration
Chemical Name or Material zwitterionic buffer
Recommended Storage 15°C to 30°C (Do not freeze).