missing translation for 'onlineSavingsMsg'
Learn More

MOPS (Fine White Crystals/Molecular Biology), Fisher BioReagents™

Commonly used buffering agent

$104.58 - $432.03

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(4-Morpholino)propane sulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP308-100
SDS View Documents Promotion Details
Fisher BioReagents
BP308100
100 g Glass Bottle
Each for $104.58
Case of 6 Each for $627.48
Only null left
 
BP308-500
SDS View Documents Promotion Details
Fisher BioReagents
BP308500
500 g Poly Bottle
Each for $432.03
Case of 6 Each for $2,592.18
Only null left
 
Description

Description

Commonly used buffering agent

This biological buffer has a usable pH range of 6.5 to 7.9.
Specifications

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
3-morpholin-4-ylpropane-1-sulfonic acid
C7H15NO4S
MFCD00006183
3-(4-Morpholino)propane sulfonic acid
CHEBI:44115
[O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

277°C
White
2.5 to 4.0
100 g
Pass Test
C7H15NO4S
MFCD00006183
Protease free
3-(4-Morpholino)propane sulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
≥97%
MOPS
1132-61-2
>277°C
Powder or Solid
DNase free
≥97 %
CH2CH2OCH2CH2N(CH2)3SO3H
Glass Bottle
15, 6352
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
209.26
DNase-, RNase- and Protease-Free
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Causes eye, skin, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:2
Flammability:1
Instability:0

Recommended Storage : RT

SDS
Documents

Documents