missing translation for 'onlineSavingsMsg'
Learn More

MOPS (Fine White Crystals/Molecular Biology), Fisher BioReagents™

Catalog No. BP308500
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP308500 500 g Poly Bottle
BP308100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP308500 Supplier Fisher BioReagents Supplier No. BP308 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Commonly used buffering agent

This biological buffer has a usable pH range of 6.5 to 7.9.

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(4-Morpholino)propane sulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Melting Point 277°C
Boiling Point 277°C
Color White
Decomposition Information >277°C
pH 2.5 to 4.0
Physical Form Crystals
Quantity 500 g
DNase DNase free
Formulation 3-(4-Morpholino) Propane Sulfonic Acid
Assay Percent Range ≥97 %
Identification Pass Test
Linear Formula CH2CH2OCH2CH2N(CH2)3SO3H
Packaging Poly Bottle
Protease Protease free
Merck Index 15, 6352
Formula Weight 209.26
Percent Purity ≥97%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material MOPS
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Causes eye, skin, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage RT