missing translation for 'onlineSavingsMsg'
Learn More

MOPS Buffer,Corning™

Catalog No. MT46103RM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT46103RM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT46103RM Supplier Corning Incorporated Supplier No. 46103RM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Color White
Physical Form Powder
Quantity 100 g
Packaging 1 x 100g
Formula Weight 209.27
Chemical Name or Material MOPS Buffer