missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 99%

Catalog No. AAA1291436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1291436 500 g
AAA1291414 25 g
AAA1291422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1291436 Supplier Thermo Scientific Chemicals Supplier No. A1291436

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.

Solubility
Soluble in water.    

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 4-Morpholinepropanesulfonic acid; 3-(4-Morpholinyl)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Melting Point 277°C to 280°C (decomposition)
Quantity 500 g
Flash Point 110°C (230°F)
Assay Percent Range 99%
Beilstein 1106776
Merck Index 14,6265
Solubility Information Soluble in water.
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
Formula Weight 209.27
Percent Purity 99%
Chemical Name or Material MOPS
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-478-5
RTECSNumber QE9104530
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only