missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MOPS, 99.5%, for molecular biology, Dnase, Rnase, Prot. free, for peptide sequencing

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

$536.40 - $1,416.49

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-Morpholino)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC327661000
SDS View Documents
Thermo Scientific Chemicals
327661000
100 g Glass bottle N/A
Only null left
 
AC327665000
SDS View Documents
Thermo Scientific Chemicals
327665000
500 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
3-morpholin-4-ylpropane-1-sulfonic acid
C7H15NO4S
MFCD00006183
3-(N-Morpholino)propanesulfonic acid
CHEBI:44115
[O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

0.01 max. at 280nm
277.0°C to 282.0°C
99.4
White
Powder
116°C
Authentic
C7H15NO4S
Glass bottle
3-(N-Morpholino)propanesulfonic acid
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
209.26
Molecular Biology
(0.1M aq. soln.)
1132-61-2
100.0
3.0 to 4.5 (1% soln. at 25°C)
100 g
99.50%
0.5% max. (105°C)
MFCD00006183
15, 6352
Solubility in water: 1000g/L (20°C). Other solubilities: <10g/L ethanol (20°C)
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
99.5%
MOPS, DNAse, RNAse and Protease Free
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

RUO – Research Use Only