missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MOPS, 99.5%, for molecular biology, Dnase, Rnase, Prot. free, for peptide sequencing

Catalog No. AC327661000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327661000 100 g Glass bottle
AC327665000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327661000 Supplier Thermo Scientific Chemicals Supplier No. 327661000

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-Morpholino)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Absorbance 0.01 max. at 280nm
Absorbance Conditions (0.1M aq. soln.)
Melting Point 277.0°C to 282.0°C
CAS Min % 99.4
Color White
CAS Max % 100.0
pH 3.0 to 4.5 (1% soln. at 25°C)
Physical Form Powder
Quantity 100 g
Flash Point 116°C
Assay Percent Range 99.50%
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (105°C)
Packaging Glass bottle
Merck Index 15, 6352
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: <10g/L ethanol (20°C)
Formula Weight 209.26
Percent Purity 99.5%
Grade Molecular Biology
Chemical Name or Material MOPS, DNAse, RNAse and Protease Free
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only