missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, ≥98.5% (Dry Basis), Ultrapure
CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
$165.37 - $646.84
Chemical Identifiers
| CAS | 1132-61-2 |
|---|---|
| Molecular Formula | C7H15NO4S |
| Molecular Weight (g/mol) | 209.26 |
| MDL Number | MFCD00006183 |
| InChI Key | DVLFYONBTKHTER-UHFFFAOYSA-N |
| Synonym | 3-[N-Morpholino]propane sulfonic Acid |
| PubChem CID | 70807 |
| ChEBI | CHEBI:44115 |
| IUPAC Name | 3-morpholin-4-ylpropane-1-sulfonic acid |
| SMILES | C1COCCN1CCCS(=O)(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AAJ1925622
|
Thermo Fisher Scientific
J1925622 |
100 g | Plastic bottle |
Each for $165.37
|
|
||||
|
AAJ19256A1
|
Thermo Fisher Scientific
J19256A1 |
1 kg | Plastic bottle |
Each for $646.84
|
|
||||
Chemical Identifiers
| 1132-61-2 | |
| 209.26 | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| 70807 | |
| 3-morpholin-4-ylpropane-1-sulfonic acid |
| C7H15NO4S | |
| MFCD00006183 | |
| 3-[N-Morpholino]propane sulfonic Acid | |
| CHEBI:44115 | |
| C1COCCN1CCCS(=O)(=O)O |
Specifications
| 277°C to 280°C (decomposition) | |
| Crystalline Powder | |
| 110°C | |
| HO3S(CH2)3(C4H8NO) | |
| Plastic bottle | |
| 3-[N-Morpholino]propane sulfonic Acid | |
| C1COCCN1CCCS(=O)(=O)O | |
| 209.26 | |
| CHEBI:44115 | |
| ≥98.5% | |
| Dry basis |
| 1132-61-2 | |
| 100 g | |
| C7H15NO4S | |
| MFCD00006183 | |
| 1106776 | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| 3-morpholin-4-ylpropane-1-sulfonic acid | |
| 70807 | |
| 209.26 | |
| MOPS |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-478-5
RTECSNumber : QE9104530
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only