missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, 101%, MP Biomedicals™
$383.34 - $383.34
Chemical Identifiers
| CAS | 71119-22-7 |
|---|---|
| Molecular Formula | C7H14NNaO4S |
| Molecular Weight (g/mol) | 231.242 |
| InChI Key | MWEMXEWFLIDTSJ-UHFFFAOYSA-M |
| Synonym | 3-[N-Morpholino]propanesulfonic acid |
| PubChem CID | 3859613 |
| IUPAC Name | sodium;3-morpholin-4-ylpropane-1-sulfonate |
| SMILES | C1COCCN1CCCS(=O)(=O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19067083
|
MP Biomedicals Inc
0219067083 |
250 g |
Each for $383.34
|
|
|||||
Chemical Identifiers
| 71119-22-7 | |
| 231.242 | |
| 3-[N-Morpholino]propanesulfonic acid | |
| sodium;3-morpholin-4-ylpropane-1-sulfonate |
| C7H14NNaO4S | |
| MWEMXEWFLIDTSJ-UHFFFAOYSA-M | |
| 3859613 | |
| C1COCCN1CCCS(=O)(=O)[O-].[Na+] |
Specifications
| 71119-22-7 | |
| Crystalline Powder | |
| 1.01 | |
| 3-[N-Morpholino]propanesulfonic acid | |
| C1COCCN1CCCS(=O)(=O)[O-].[Na+] | |
| 231.242 | |
| 231.2 |
| White | |
| 250 g | |
| C7H14NNaO4S | |
| MWEMXEWFLIDTSJ-UHFFFAOYSA-M | |
| sodium;3-morpholin-4-ylpropane-1-sulfonate | |
| 3859613 | |
| MOPS |