missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, 1.0M buffer soln., pH 9.0, Thermo Scientific™
$148.15 - $148.15
Chemical Identifiers
| CAS | 1132-61-2 |
|---|---|
| Molecular Formula | C7H15NO4S |
| Molecular Weight (g/mol) | 209.26 |
| MDL Number | MFCD00006183 |
| InChI Key | DVLFYONBTKHTER-UHFFFAOYSA-N |
| PubChem CID | 70807 |
| ChEBI | CHEBI:44115 |
| IUPAC Name | 3-morpholin-4-ylpropane-1-sulfonic acid |
| SMILES | [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ60097AK
|
Thermo Scientific Chemicals
J60097AK |
250 mL |
Each for $148.15
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1132-61-2 | |
| 209.26 | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| CHEBI:44115 | |
| [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
| C7H15NO4S | |
| MFCD00006183 | |
| 70807 | |
| 3-morpholin-4-ylpropane-1-sulfonic acid |
Specifications
| 1132-61-2 | |
| 1.0 M | |
| Liquid | |
| C7H15NO4S | |
| MFCD00006183 | |
| [O-]S(=O)(=O)CCC[NH+]1CCOCC1 | |
| 209.26 | |
| CHEBI:44115 |
| Colorless | |
| 9.0 | |
| 250 mL | |
| Odorless | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| 3-morpholin-4-ylpropane-1-sulfonic acid | |
| 70807 | |
| MOPS buffer soln. |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures