missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MOPS, 1.0M buffer soln., pH 6.5

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

Supplier:  Thermo Scientific Chemicals J61797AP

 View more versions of this product

Catalog No. AAJ61797AP


Only null left

Description

Description

MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a morpholino propanesulfonic acid buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a morpholino propanesulfonic acid buffer.

Solubility
Soluble in water.

Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
Specifications

Specifications

MOPS buffer soln.
Colorless
6.5
500 mL
Odorless
Soluble in water.
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
1132-61-2
1.0 M
Liquid
C7H15NO4S
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
CHEBI:44115
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
C7H15NO4S
MFCD00006183
70807
3-morpholin-4-ylpropane-1-sulfonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only