missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, 1.0M buffer soln., pH 5.5, Thermo Scientific™
$210.66 - $280.29
Chemical Identifiers
| CAS | 1132-61-2 |
|---|---|
| Molecular Formula | C7H15NO4S |
| Molecular Weight (g/mol) | 209.26 |
| MDL Number | MFCD00006183 |
| InChI Key | DVLFYONBTKHTER-UHFFFAOYSA-N |
| PubChem CID | 70807 |
| ChEBI | CHEBI:44115 |
| IUPAC Name | 3-morpholin-4-ylpropane-1-sulfonic acid |
| SMILES | [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ62261AK
|
Thermo Scientific Chemicals
J62261AK |
250 mL |
Each for $210.66
|
|
|||||
|
AAJ62261AP
|
Thermo Scientific Chemicals
J62261AP |
500 mL |
Each for $280.29
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsMOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a morpholino propanesulfonic acid buffer.
Solubility
Soluble in water.
Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
Chemical Identifiers
| 1132-61-2 | |
| 209.26 | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| CHEBI:44115 | |
| [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
| C7H15NO4S | |
| MFCD00006183 | |
| 70807 | |
| 3-morpholin-4-ylpropane-1-sulfonic acid |
Specifications
| 1132-61-2 | |
| 1.0 M | |
| Liquid | |
| C7H15NO4S | |
| MFCD00006183 | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| 3-morpholin-4-ylpropane-1-sulfonic acid | |
| 70807 | |
| MOPS buffer soln. |
| Colorless | |
| 5.5 | |
| 250 mL | |
| Odorless | |
| Soluble in water. | |
| [O-]S(=O)(=O)CCC[NH+]1CCOCC1 | |
| 209.26 | |
| CHEBI:44115 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Keep cold