missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MOPS, 0.5M buffer soln., pH 7.5

Catalog No. AAJ62839AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62839AE 100 mL
AAJ62839AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62839AE Supplier Thermo Scientific Chemicals Supplier No. J62839AE

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

Solubility
Soluble in water.

Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Concentration 0.5 M
pH 7.5
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Solubility Information Soluble in water.
Chemical Name or Material MOPS buffer soln.
Hazard Category H315-H319-H335
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only