missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals MOPS, 0.5M buffer soln., pH 7.0
CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
$150.07 - $222.84
Chemical Identifiers
| CAS | 1132-61-2 |
|---|---|
| Molecular Formula | C7H15NO4S |
| Molecular Weight (g/mol) | 209.26 |
| MDL Number | MFCD00006183 |
| InChI Key | DVLFYONBTKHTER-UHFFFAOYSA-N |
| PubChem CID | 70807 |
| ChEBI | CHEBI:44115 |
| IUPAC Name | 3-morpholin-4-ylpropane-1-sulfonic acid |
| SMILES | [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ60328AE
|
Thermo Scientific Chemicals
J60328AE |
100 mL |
Each for $150.07
|
|
|||||
|
AAJ60328AK
|
Thermo Scientific Chemicals
J60328AK |
250 mL |
Each for $222.84
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1132-61-2 | |
| 209.26 | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| CHEBI:44115 | |
| [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
| C7H15NO4S | |
| MFCD00006183 | |
| 70807 | |
| 3-morpholin-4-ylpropane-1-sulfonic acid |
Specifications
| 1132-61-2 | |
| 7.0 | |
| 100 mL | |
| Odorless | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| 3-morpholin-4-ylpropane-1-sulfonic acid | |
| 70807 | |
| MOPS buffer soln. |
| 0.5 M | |
| Liquid | |
| C7H15NO4S | |
| MFCD00006183 | |
| [O-]S(=O)(=O)CCC[NH+]1CCOCC1 | |
| 209.26 | |
| CHEBI:44115 |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only