missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monomethyl Itaconate 98.0+%, TCI America™
$177.08 - $1399.90
Chemical Identifiers
| CAS | 7338-27-4 |
|---|---|
| Molecular Formula | C6H8O4 |
| Molecular Weight (g/mol) | 144.126 |
| MDL Number | MFCD00021713 |
| InChI Key | OIYTYGOUZOARSH-UHFFFAOYSA-N |
| Synonym | Itaconic Acid Monomethyl Ester, beta-Methyl Itaconate, 4-Methyl Methylenesuccinate |
| PubChem CID | 81791 |
| IUPAC Name | 4-methoxy-2-methylidene-4-oxobutanoic acid |
| SMILES | COC(=O)CC(=C)C(=O)O |
Chemical Identifiers
| 7338-27-4 | |
| 144.126 | |
| OIYTYGOUZOARSH-UHFFFAOYSA-N | |
| 81791 | |
| COC(=O)CC(=C)C(=O)O |
| C6H8O4 | |
| MFCD00021713 | |
| Itaconic Acid Monomethyl Ester, beta-Methyl Itaconate, 4-Methyl Methylenesuccinate | |
| 4-methoxy-2-methylidene-4-oxobutanoic acid |
Specifications
| 7338-27-4 | |
| White | |
| C6H8O4 | |
| 25 g | |
| OIYTYGOUZOARSH-UHFFFAOYSA-N | |
| 4-methoxy-2-methylidene-4-oxobutanoic acid | |
| 81791 | |
| ≥98.0% (GC,T) | |
| Monomethyl Itaconate |
| 72°C | |
| 149°C | |
| MFCD00021713 | |
| Itaconic Acid Monomethyl Ester, beta-Methyl Itaconate, 4-Methyl Methylenesuccinate | |
| COC(=O)CC(=C)C(=O)O | |
| 144.126 | |
| 144.13 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-2615
TSCA : No
Recommended Storage : Refrigerator