missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monomethyl Itaconate 98.0+%, TCI America™
Supplier: TCI America I0269500G
Specifications
| Monomethyl Itaconate | |
| 72°C | |
| 149°C | |
| MFCD00021713 | |
| Itaconic Acid Monomethyl Ester, beta-Methyl Itaconate, 4-Methyl Methylenesuccinate | |
| COC(=O)CC(=C)C(=O)O | |
| 144.126 | |
| 144.13 | |
| Crystalline Powder |
| 7338-27-4 | |
| White | |
| C6H8O4 | |
| 500 g | |
| OIYTYGOUZOARSH-UHFFFAOYSA-N | |
| 4-methoxy-2-methylidene-4-oxobutanoic acid | |
| 81791 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 7338-27-4 | |
| 144.126 | |
| OIYTYGOUZOARSH-UHFFFAOYSA-N | |
| 81791 | |
| COC(=O)CC(=C)C(=O)O |
| C6H8O4 | |
| MFCD00021713 | |
| Itaconic Acid Monomethyl Ester, beta-Methyl Itaconate, 4-Methyl Methylenesuccinate | |
| 4-methoxy-2-methylidene-4-oxobutanoic acid |
Safety and Handling
EINECSNumber : (2)-2615
TSCA : No
Recommended Storage : Refrigerator