missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Isophthalate 97.0+%, TCI America™

Catalog No. M23271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23271G 1 g
M23275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23271G Supplier TCI America Supplier No. M23271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1877-71-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00029972
InChI Key WMZNGTSLFSJHMZ-UHFFFAOYSA-N
Synonym 3-methoxycarbonyl benzoic acid, mono-methyl isophthalate, methyl hydrogen isophthalate, isophthalic acid monomethyl ester, monomethyl isophthalate, isophthalic acid methyl ester, isophthalic acid, methyl ester, isophthalic acid, monomethyl ester, methyl m-phthalate, methyl 3-carboxybenzoate
PubChem CID 601880
IUPAC Name 3-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 1 g
Formula Weight 180.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl Isophthalate