missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl 5-Nitroisophthalate 98.0+%, TCI America™

Catalog No. N064125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N064125G 25 g
N0641500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N064125G Supplier TCI America Supplier No. N064125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1955-46-0
Molecular Formula C9H7NO6
Molecular Weight (g/mol) 225.156
MDL Number MFCD00009793
InChI Key ZCRNIIJXDRYWDU-UHFFFAOYSA-N
Synonym 3-methoxycarbonyl-5-nitrobenzoic acid, 5-nitroisophthalic acid monomethyl ester, monomethyl 5-nitroisophthalate, methyl 5-nitroisophthalate, mono-methyl 5-nitroisophthalate, mono-methyl-5-nitroisophthalic acid, methyl hydrogen 5-nitroisophthalate, 5-nitroisophthalic acid, monomethyl ester, monomethyl-5-nitroisophthalate, monomethyl 5-nitrobenzene-1,3-dicarboxylate
PubChem CID 74776
SMILES COC(=O)C1=CC(=CC(=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 25 g
Formula Weight 225.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl 5-Nitroisophthalate