missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monomagnesium EDTA, 0.2 g/L, for Hardness, Ricca Chemical
$112.00 - $112.00
Chemical Identifiers
| CAS | 14402-88-1 |
|---|---|
| Molecular Formula | C10H12MgN2Na2O8 |
| Molecular Weight (g/mol) | 358.50 |
| MDL Number | MFCD00078217 |
| InChI Key | AWNVVAMWLMUZOZ-UHFFFAOYSA-J |
| Synonym | unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate |
| PubChem CID | 161064 |
| IUPAC Name | magnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate |
| SMILES | [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O |
Chemical Identifiers
| 14402-88-1 | |
| 358.50 | |
| AWNVVAMWLMUZOZ-UHFFFAOYSA-J | |
| 161064 | |
| [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O |
| C10H12MgN2Na2O8 | |
| MFCD00078217 | |
| unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate | |
| magnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate |
Specifications
| Colorless | |
| 98.0,0.02 | |
| C10H12MgN2Na2O8 | |
| unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate | |
| [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O | |
| 358.50 | |
| Laboratory | |
| 500 mL | |
| 0.2g/L |
| 14402-88-1 , 7732-18-5 | |
| 100.0,0.02 | |
| MFCD00078217 | |
| AWNVVAMWLMUZOZ-UHFFFAOYSA-J | |
| magnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate | |
| 161064 | |
| Natural Poly Bottle | |
| Liquid | |
| Monomagnesium EDTA |