missing translation for 'onlineSavingsMsg'
Learn More

Monoethyl 5-Nitroisophthalate 98.0+%, TCI America™

Catalog No. N0676500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0676500G 500 g
N067625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0676500G Supplier TCI America Supplier No. N0676500G
Only null left

Chemical Identifiers

CAS 22871-55-2
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.183
MDL Number MFCD00130051
InChI Key HOYHBDFTELAGGG-UHFFFAOYSA-N
Synonym 5-Nitroisophthalic Acid Monoethyl Ester
PubChem CID 89872
IUPAC Name 3-ethoxycarbonyl-5-nitrobenzoic acid
SMILES CCOC(=O)C1=CC(=CC(=C1)C(=O)O)[N+](=O)[O-]

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 239.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monoethyl 5-Nitroisophthalate