accessibility menu, dialog, popup

UserName

CH3OONOOOHO

CAS RN 22871-55-2

IUPAC Name: 3-(ethoxycarbonyl)-5-nitrobenzoic acid
Synonyms: 3-(ethoxycarbonyl)-5-nitrobenzoic acid
Molecular Weight (g/mol): 239.18
Molecular Formula: C10H9NO6
InChi Key: HOYHBDFTELAGGG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results