missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monobasic Potassium Phosphate, Crystal, NF, 98-100.5%, Spectrum™ Chemical
PO200, 7778-77-0, KH2PO4
$1741.30 - $8840.07
Chemical Identifiers
| CAS | 7778-77-0 |
|---|---|
| Molecular Formula | H2KO4P |
| Molecular Weight (g/mol) | 136.08 |
| MDL Number | MFCD00011401 MFCD00147253 |
| InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
| IUPAC Name | potassium dihydrogen phosphate |
| SMILES | [K+].OP(O)([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604653
|
Spectrum Chemical Mfg Cor
PO20025KG |
2.5 kg | Poly Bottle |
Each for $1,741.30
|
|
||||
|
18604652
|
Spectrum Chemical Mfg Cor
PO200500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $2,268.60
|
|
||||
|
18604654
|
Spectrum Chemical Mfg Cor
PO20025KGCS4 |
4 x 2.5 kg | Poly Bottle |
Case of 4 Each for $5,813.33
|
|
||||
|
18604655
|
Spectrum Chemical Mfg Cor
PO20012KG |
12 kg | Poly Pail |
Each for $4,087.07
|
|
||||
|
18604656
|
Spectrum Chemical Mfg Cor
PO20045KGBL |
45 kg | Fiber Drum |
Each for $8,840.07
|
|
||||
Description
Spectrum™ Chemical Monobasic Potassium Phosphate, Crystal, NF is used as a buffering agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 7778-77-0 | |
| 136.08 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| [K+].OP(O)([O-])=O |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate |
Specifications
| 7778-77-0 | |
| 2.5 kg | |
| MFCD00011401 MFCD00147253 | |
| [K+].OP(O)([O-])=O | |
| 136.08 | |
| NF | |
| Poly Bottle |
| 1 | |
| H2KO4P | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| potassium dihydrogen phosphate | |
| 98 to 100.5% | |
| 0.01 |