missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monobasic Potassium Phosphate, Crystal, NF, 98-100.5%, Spectrum™ Chemical
PO200, 7778-77-0, KH2PO4
Supplier: Spectrum Chemical Mfg Cor PO20012KG
Description
Spectrum™ Chemical Monobasic Potassium Phosphate, Crystal, NF is used as a buffering agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 7778-77-0 | |
| 12 kg | |
| MFCD00011401 MFCD00147253 | |
| [K+].OP(O)([O-])=O | |
| 136.08 | |
| NF | |
| Poly Pail |
| 1 | |
| H2KO4P | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| potassium dihydrogen phosphate | |
| 98 to 100.5% | |
| 0.01 |
Chemical Identifiers
| 7778-77-0 | |
| 136.08 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| [K+].OP(O)([O-])=O |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate |