missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum AA Standard, 1000 ppm Mo in 3% NH4OH, Ricca Chemical

$290.16 - $290.16

Chemical Identifiers

CAS 13106-76-8
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AMO1KW-500
View Documents
Ricca Chemical Company
AMO1KW500
500 mL Natural Poly Bottle
Each for $290.16
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13106-76-8

Specifications

Natural Poly Bottle
95.0,2.85,0.19
99.0,2.96,0.2
Liquid
100°C
Molybdenum AA Standard
13106-76-8,1336-21-6,13106-76-8,7732-18-5
1,000ppm Mo
1 mL = 1 mg Mo
Colorless to Green/Yellow
500 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'