missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum AA Standard, 1000 ppm Mo in 3% HNO3, Ricca Chemical
$112.47 - $112.47
Chemical Identifiers
| 13106-76-8 |
Specifications
| Natural LDPE Bottle | |
| 94.0,4.22,0.19 | |
| Liquid | |
| < 2 | |
| Standard | |
| 1mL = 1mg Mo (1, 000ppm Mo)(NH4)2MoO4 in 3% HNO3 |
| 13106-76-8,7697-37-2,13106-76-8,7732-18-5 | |
| 97.0,4.39,0.2 | |
| Colorless | |
| Approximately 0°C | |
| 100 mL | |
| Molybdenum AA Standard |
Safety and Handling
ShelfLife : 24 months