missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum AA Standard, 1000 ppm Mo in 3% HCl/tr HF, Ricca Chemical
$316.77 - $316.77
Chemical Identifiers
| CAS | 1313-27-5 |
|---|---|
| Molecular Formula | MoO3 |
| Molecular Weight (g/mol) | 143.95 |
| MDL Number | MFCD00003469 |
| InChI Key | JKQOBWVOAYFWKG-UHFFFAOYSA-N |
| PubChem CID | 14802 |
| ChEBI | CHEBI:30627 |
| IUPAC Name | trioxomolybdenum |
| SMILES | O=[Mo](=O)=O |
Chemical Identifiers
| 1313-27-5 | |
| 143.95 | |
| JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
| CHEBI:30627 | |
| O=[Mo](=O)=O |
| MoO3 | |
| MFCD00003469 | |
| 14802 | |
| trioxomolybdenum |
Specifications
| Natural Poly Bottle | |
| 1,000ppm | |
| 98.0,4.06,0.15,0.01 | |
| MoO3 | |
| Colorless | |
| JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
| 3% HCl/tr HF | |
| 143.95 | |
| 14802 | |
| Molybdenum AA Standard |
| 1313-27-5,7647-01-0,1313-27-5,7664-39-3,7732-18-5 | |
| 94.0,3.9,0.14,0.01 | |
| Liquid | |
| MFCD00003469 | |
| 100°C | |
| O=[Mo](=O)=O | |
| trioxomolybdenum | |
| 500 mL | |
| CHEBI:30627 |