missing translation for 'onlineSavingsMsg'
Learn More

Molybdate Reagent, Taylor Type, 1.2% (w/v) Sodium Molybdate in 4.9% (v/v) Sulfuric Acid, Ricca Chemical

Catalog No. p-3251954
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
519032 1 L Natural Poly Bottle
51901 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 519032 Supplier Ricca Chemical Company Supplier No. 519032
Only null left

Chemical Identifiers

CAS 10102-40-6 , 7732-18-5 , 7664-93-9
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Color Colorless to Green-Yellow
CAS Min % 1.07, 89.0, 8.02
CAS Max % 1.11, 93.0, 8.34
Percent Purity 1.2%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix 4.9% (v/v) Sulfuric Acid
pH <1
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 1.2% (w/v) Sodium Molybdate
Chemical Name or Material Molybdate Reagent, Taylor Type
Show More Show Less