missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 4A, Cylinders, 3.2mm diameter

Catalog No. AC436730010
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass Bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436730010 1 kg Glass Bottle
AC436730050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436730010 Supplier Thermo Scientific Chemicals Supplier No. 436730010

CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Color Beige
Packaging Glass Bottle
Quantity 1 kg
Bulk Density 650 to 750g/L
Residual Water < 2% (900°C, 2 hrs.)
Water Adsorption > 20% (55% rH, 20°C)
Physical Form Cylinders
IUPAC Name Molecular sieves
Chemical Name or Material Molecular sieves 4A

RUO – Research Use Only