missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 4A, -8+12 (ca 2mm) beads

CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol

Supplier:  Thermo Scientific Chemicals L054540I

 View more versions of this product

Catalog No. AAL054540I

Only null left
Explore available promotions

Description

Description

Molecular sieves, 4 Å is used to absorb molecules as large as butane. It is utilized for the separation of natural gas, liquified petroleum gas (LPG) and alkenes. It finds application in the refining and purification of liquid and gases. It is also used for drying and purification of air, inert gases and solvents. It is actively involved to remove methanol, carbon dioxide and ammonia from fluid streams. It plays a vital role to remove heavy metal ions available in water. It is also used as a soap forming agent and as a detergent auxiliary. It has been considered as a general-purpose drying agent in polar and non polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5000 g
Molecular sieves, 4A
70955-01-0
MFCD00131613
FJUZEZRZKVMAMD-UHFFFAOYNA-N
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
73906282
Hygroscopic
1 to 2mm (0.04 to 0.08 in) beads
C20H25FN2O8
Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone.
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
440.42

Chemical Identifiers

70955-01-0
440.42
FJUZEZRZKVMAMD-UHFFFAOYNA-N
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
C20H25FN2O8
MFCD00131613
73906282
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only